Assay Details
Target Gene Details
Entrez Gene ID: | 7391 |
Gene Name: | upstream transcription factor 1 |
Gene Aliases: |
FCHL, FCHL1, HYPLIP1, MLTF, MLTFI, UEF, bHLHb11 |
Location: |
Chr.1:161039251-161045979 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USF1 | NM_001276373.1 | NP_001263302.1 | ||
| NM_007122.4 | NP_009053.1 | |||
| NM_207005.2 | NP_996888.1 | |||
| AB098540.1 | BAC78384.1 | |||
| AK314876.1 | ||||
| AL832119.1 | ||||
| BC035505.2 | AAH35505.1 | |||
| BP280296.1 | ||||
| KU178379.1 | ||||
| KU178380.1 | ||||
| X55666.1 | CAA39201.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3221 | Chr.1:161038385 - 161083189 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map