Assay Details
Target Gene Details
Entrez Gene ID: | 79654 |
Gene Name: | HECT domain E3 ubiquitin protein ligase 3 |
Gene Aliases: |
- |
Location: |
Chr.1:45002548-45011355 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HECTD3 | NM_024602.5 | 21 | 3304 | NP_078878.3 |
| XM_011542140.2 | 19 | 2701 | XP_011540442.1 | |
| AA001245.1 | ||||
| AK056545.1 | 19 | 2655 | ||
| AK056873.1 | 13 | 2198 | ||
| AK091583.1 | 6 | 2053 | ||
| AK096462.1 | 21 | 3183 | ||
| BC019105.2 | 6 | 1263 | ||
| CA314393.1 | 1 | 349 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950706 | Chr.1:45001329 - 45011128 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map