Assay Details
Target Gene Details
Entrez Gene ID: | 100287944 |
Gene Name: | uncharacterized LOC100287944 |
Gene Aliases: |
- |
Location: |
Chr.12:106496410-106774831 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100287944 | NR_040246.1 | |||
| DA250223.1 |
Target Gene Details
Entrez Gene ID: | 100505978 |
Gene Name: | uncharacterized LOC100505978 |
Gene Aliases: |
- |
Location: |
Chr.12:106680758-106684700 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100505978 | NR_038912.1 | 3 | 408 |
Target Gene Details
Entrez Gene ID: | 5992 |
Gene Name: | regulatory factor X4 |
Gene Aliases: |
NYD-SP10 |
Location: |
Chr.12:106582907-106762804 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RFX4 | NM_001206691.1 | NP_001193620.1 | ||
| NM_213594.2 | NP_998759.1 | |||
| AB044245.1 | BAB59001.1 | |||
| AB095365.1 | BAE48231.1 | |||
| AB095366.1 | BAE48232.1 | |||
| AK074913.1 | BAC11288.1 | |||
| AK291445.1 | ||||
| AK302837.1 | ||||
| AY102009.1 | AAM52484.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832511 | Chr.12:106551876 - 106714050 on Build GRCh38 | Gain |
|
| nsv9005 | Chr.12:106625455 - 106973352 on Build GRCh38 | Gain |
|
| nsv560090 | Chr.12:106541358 - 106830191 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map