Assay Details
Target Gene Details
Entrez Gene ID: | 139562 |
Gene Name: | OTU deubiquitinase 6A |
Gene Aliases: |
DUBA2, HSHIN6 |
Location: |
Chr.X:70062491-70064179 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OTUD6A | NM_207320.2 | 1 | 1196 | NP_997203.1 |
| AK098697.1 | 1 | 1196 | BAC05384.1 | |
| BC137355.1 | 1 | 1192 | ||
| BC137356.1 | 1 | 1192 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6949 | Chr.X:70037194 - 70070943 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map