Assay Details
Target Gene Details
Entrez Gene ID: | 1407 |
Gene Name: | cryptochrome circadian clock 1 |
Gene Aliases: |
PHLL1 |
Location: |
Chr.12:106991364-107093872 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRY1 | NM_004075.4 | 7 | 1767 | NP_004066.1 |
| XM_017018832.1 | 8 | 1902 | XP_016874321.1 | |
| AK098615.1 | 2 | 170 | BAC05354.1 | |
| AK125915.1 | 9 | 1430 | ||
| AK290552.1 | 7 | 1067 | ||
| BC030519.1 | 7 | 1440 | AAH30519.1 | |
| D83702.1 | 7 | 1457 | BAA12068.1 | |
| D84657.1 | 7 | 871 | BAA12710.1 |
More Information
Set Membership: |
|

Genomic Map