Assay Details
Target Gene Details
Entrez Gene ID: | 3490 |
Gene Name: | insulin like growth factor binding protein 7 |
Gene Aliases: |
AGM, FSTL2, IBP-7, IGFBP-7, IGFBP-7v, IGFBPRP1, MAC25, PSF, RAMSVPS, TAF |
Location: |
Chr.4:57031071-57110385 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGFBP7 | NM_001253835.1 | NP_001240764.1 | ||
| NM_001553.2 | NP_001544.1 | |||
| AF540057.1 | AAN23122.1 | |||
| AK303915.1 | ||||
| AK316082.1 | ||||
| BC017201.2 | AAH17201.1 | |||
| BC066339.1 | AAH66339.1 | |||
| BT006654.1 | AAP35300.1 | |||
| L19182.1 | AAA16187.1 | |||
| S75725.1 | AAB32370.1 |
Target Gene Details
Entrez Gene ID: | 255130 |
Gene Name: | IGFBP7 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.4:57109762-57205299 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGFBP7-AS1 | NR_034081.1 | 1 | 128 | |
| BG425784.1 |
More Information
Set Membership: |
|

Genomic Map