Assay Details
Target Gene Details
Entrez Gene ID: | 414157 |
Gene Name: | chromosome 10 open reading frame 62 |
Gene Aliases: |
bA548K23.1 |
Location: |
Chr.10:97589693-97590934 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C10orf62 | NM_001009997.2 | 1 | 852 | NP_001009997.2 |
| BC037252.1 | 1 | 818 | AAH37252.1 |
Target Gene Details
Entrez Gene ID: | 112817 |
Gene Name: | 4-hydroxy-2-oxoglutarate aldolase 1 |
Gene Aliases: |
C10orf65, DHDPS2, DHDPSL, HP3, NPL2 |
Location: |
Chr.10:97584345-97612802 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HOGA1 | NM_001134670.1 | NP_001128142.1 | ||
| NM_138413.3 | NP_612422.2 | |||
| AK094791.1 | ||||
| AK289440.1 | ||||
| BC011916.2 | AAH11916.1 | |||
| BC045550.1 | AAH45550.1 | |||
| BC057821.1 | AAH57821.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1047359 | Chr.10:97546744 - 97609256 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map