Assay Details
Target Gene Details
Entrez Gene ID: | 7957 |
Gene Name: | epilepsy, progressive myoclonus type 2A, Lafora disease (laforin) |
Gene Aliases: |
EPM2, MELF |
Location: |
Chr.6:145500744-145736018 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPM2A | NM_001018041.1 | NP_001018051.1 | ||
| NM_005670.3 | NP_005661.1 | |||
| XM_006715564.3 | 1 | 186 | XP_006715627.1 | |
| XM_011536113.2 | 1 | 186 | XP_011534415.1 | |
| AF284580.1 | AAG18377.1 | |||
| AF454491.1 | AAO15523.1 | |||
| AK299497.1 | ||||
| AW291546.1 | 1 | 264 |
Target Gene Details
Entrez Gene ID: | 100507557 |
Gene Name: | uncharacterized LOC100507557 |
Gene Aliases: |
- |
Location: |
Chr.6:145734869-145886585 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100507557 | NR_038246.1 | |||
| AK124757.1 | ||||
| DB465589.1 |
More Information
Set Membership: |
|

Genomic Map