Assay Details
Target Gene Details
Entrez Gene ID: | 65989 |
Gene Name: | delta like non-canonical Notch ligand 2 |
Gene Aliases: |
DLK-2, EGFL9 |
Location: |
Chr.6:43450352-43456632 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DLK2 | NM_001286655.1 | NP_001273584.1 | ||
| NM_001286656.1 | NP_001273585.1 | |||
| NM_023932.3 | NP_076421.2 | |||
| NM_206539.2 | NP_996262.1 | |||
| XM_005249308.4 | XP_005249365.1 | |||
| XM_011514823.2 | XP_011513125.1 | |||
| XM_017011204.1 | 1 | 486 | XP_016866693.1 | |
| AK055380.1 | ||||
| AY358126.1 | AAQ88493.1 | |||
| BC000230.1 | AAH00230.1 | |||
| BC006425.1 | AAH06425.1 | |||
| BC110320.1 | AAI10321.1 | |||
| BI916137.1 | ||||
| DA332624.1 | ||||
| HY064089.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv603000 | Chr.6:43397622 - 43524840 on Build GRCh38 | Loss |
|
| nsv950172 | Chr.6:43440263 - 43458862 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map