Assay Details
Target Gene Details
Entrez Gene ID: | 2618 |
Gene Name: | phosphoribosylglycinamide formyltransferase, phosphoribosylglycinamide synthetase, phosphoribosylaminoimidazole synthetase |
Gene Aliases: |
AIRS, GARS, GARTF, PAIS, PGFT, PRGS |
Location: |
Chr.21:33503931-33542917 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GART | NM_000819.4 | 22 | 3146 | NP_000810.1 |
| NM_001136005.1 | 22 | 3322 | NP_001129477.1 | |
| NM_001136006.1 | 22 | 3285 | NP_001129478.1 | |
| XM_005260941.1 | 22 | 3422 | XP_005260998.1 | |
| XM_006723989.1 | 22 | 3364 | XP_006724052.1 | |
| XM_006723990.1 | 22 | 3252 | XP_006724053.1 | |
| XM_011529526.1 | 22 | 3331 | XP_011527828.1 | |
| AB208785.1 | 22 | 3212 | BAD92022.1 | |
| AK292560.1 | 22 | 3261 | ||
| AK292897.1 | 22 | 3099 | ||
| AK295980.1 | 12 | 1766 | ||
| BC107712.1 | 22 | 3100 | AAI07713.1 | |
| BQ010035.1 | 1 | 231 | ||
| BX648124.1 | 22 | 3263 | ||
| M32082.1 | 6 | 887 | AAA60077.1 | |
| X54199.1 | 22 | 3089 | CAA38119.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509795 | Chr.21:33457276 - 33521074 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map