Assay Details
Target Gene Details
Entrez Gene ID: | 131177 |
Gene Name: | family with sequence similarity 3 member D |
Gene Aliases: |
EF7, OIT1 |
Location: |
Chr.3:58633943-58666843 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM3D | NM_138805.2 | 9 | 822 | NP_620160.1 |
| XM_005264862.4 | 9 | 795 | XP_005264919.1 | |
| XM_005264863.4 | 9 | 828 | XP_005264920.1 | |
| XM_005264864.4 | 9 | 800 | XP_005264921.1 | |
| XM_006712965.3 | 9 | 592 | XP_006713028.1 | |
| XM_006712967.3 | 8 | 719 | XP_006713030.1 | |
| XM_011533348.2 | 9 | 849 | XP_011531650.1 | |
| XM_011533349.2 | 9 | 778 | XP_011531651.1 | |
| XM_011533350.2 | 9 | 846 | XP_011531652.1 | |
| XM_011533351.2 | 9 | 868 | XP_011531653.1 | |
| XM_011533352.2 | 8 | 722 | XP_011531654.1 | |
| XM_011533353.2 | 8 | 652 | XP_011531655.1 | |
| XM_011533354.2 | 8 | 649 | XP_011531656.1 | |
| AB105189.1 | 9 | 825 | ||
| AF494381.1 | 7 | 525 | AAM94282.1 | |
| AK127546.1 | 9 | 817 | ||
| AW662283.1 | ||||
| AY358677.1 | 9 | 836 | AAQ89040.1 | |
| BC015359.1 | 9 | 731 | AAH15359.1 | |
| CA314533.1 | 2 | 506 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4749n100 | Chr.3:58504269 - 60219035 on Build GRCh38 | Gain |
|
| nsv590379 | Chr.3:58628289 - 58656725 on Build GRCh38 | Loss |
|
| esv3596253 | Chr.3:58621679 - 58677821 on Build GRCh38 | Gain |
|
| esv3893702 | Chr.3:58557785 - 58858605 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map