Assay Details
Target Gene Details
Entrez Gene ID: | 214 |
Gene Name: | activated leukocyte cell adhesion molecule |
Gene Aliases: |
CD166, MEMD |
Location: |
Chr.3:105366713-105576913 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALCAM | NM_001243280.1 | 1 | 376 | NP_001230209.1 |
| NM_001243281.1 | 1 | 376 | NP_001230210.1 | |
| NM_001243283.1 | 1 | 376 | NP_001230212.1 | |
| NM_001627.3 | 1 | 376 | NP_001618.2 | |
| AK054632.1 | 1 | 329 | ||
| AK316113.1 | 1 | 180 | ||
| AL833702.1 | 1 | 177 | ||
| AY644765.1 | AAV28819.1 | |||
| BC057809.1 | 1 | 180 | AAH57809.1 | |
| CB145726.1 | 1 | 220 | ||
| DA318514.1 | 1 | 376 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525071 | Chr.3:105286009 - 105690477 on Build GRCh38 | Gain |
|
| nsv829650 | Chr.3:105352049 - 105536192 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map