Assay Details
Target Gene Details
Entrez Gene ID: | 7066 |
Gene Name: | thrombopoietin |
Gene Aliases: |
MGDF, MKCSF, ML, MPLLG, THCYT1, TPO |
Location: |
Chr.3:184371935-184379688 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THPO | NM_000460.3 | 6 | 1055 | NP_000451.1 |
| NM_001177597.2 | 6 | 1043 | NP_001171068.1 | |
| NM_001177598.2 | 6 | 1038 | NP_001171069.1 | |
| NM_001289997.1 | 7 | 939 | NP_001276926.1 | |
| NM_001289998.1 | 7 | 1322 | NP_001276927.1 | |
| NM_001290003.1 | 7 | 1322 | NP_001276932.1 | |
| NM_001290022.1 | 7 | 1310 | NP_001276951.1 | |
| NM_001290026.1 | 7 | 1305 | NP_001276955.1 | |
| NM_001290027.1 | 8 | 1206 | NP_001276956.1 | |
| NM_001290028.1 | 6 | 1019 | NP_001276957.1 | |
| XM_017007107.1 | 8 | 1194 | XP_016862596.1 | |
| AB102890.1 | 4 | 759 | BAD89419.1 | |
| AK311002.1 | 4 | 449 | ||
| BC130322.1 | 6 | 975 | ||
| BC136338.1 | 6 | 987 | ||
| BC143982.1 | 6 | 975 | ||
| D32047.1 | 5 | 877 | BAA21930.1 | |
| L33410.1 | 6 | 991 | AAA59857.1 | |
| L36052.1 | 4 | 776 | AAC37566.1 | |
| U11025.1 | 5 | 811 | AAA50553.1 | |
| U42977.1 | ||||
| U59493.1 | 4 | 776 | AAB03392.1 | |
| U59494.1 | 4 | 776 | AAB03393.1 | |
| U59495.1 | 5 | 659 | AAB03394.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422400 | Chr.3:184299495 - 184425713 on Build GRCh38 | Duplication |
|
| nsv829813 | Chr.3:184264997 - 184427996 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map