Assay Details
Target Gene Details
Entrez Gene ID: | 101926913 |
Gene Name: | uncharacterized LOC101926913 |
Gene Aliases: |
- |
Location: |
Chr.2:170700368-170770766 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101926913 | NR_110185.1 | |||
| BM931568.1 |
Target Gene Details
Entrez Gene ID: | 389058 |
Gene Name: | Sp5 transcription factor |
Gene Aliases: |
- |
Location: |
Chr.2:170714725-170717988 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SP5 | NM_001003845.2 | 2 | 1889 | NP_001003845.1 |
| XM_005246542.4 | 2 | 3016 | XP_005246599.1 | |
| XM_011511159.2 | 2 | 3052 | XP_011509461.1 | |
| AB096175.1 | 2 | 1885 | BAD34944.1 | |
| AW450640.1 | 1 | 75 | ||
| BQ064677.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv821786 | Chr.2:170711270 - 170718197 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map