Assay Details
Target Gene Details
Entrez Gene ID: | 7023 |
Gene Name: | transcription factor AP-4 (activating enhancer binding protein 4) |
Gene Aliases: |
AP-4, bHLHc41 |
Location: |
Chr.16:4257186-4273000 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TFAP4 | NM_003223.2 | NP_003214.1 | ||
| XM_011522633.2 | XP_011520935.1 | |||
| XM_011522635.2 | XP_011520937.1 | |||
| AB209146.1 | BAD92383.1 | |||
| BC010576.1 | AAH10576.1 | |||
| BU159415.1 | ||||
| CR541726.1 | CAG46527.1 | |||
| S73885.1 | AAB32235.1 | |||
| X57435.1 | CAA40683.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv21490 | Chr.16:3825338 - 4270692 on Build GRCh38 | Gain+Loss |
|
| nsv1046986 | Chr.16:4049000 - 5555721 on Build GRCh38 | Gain |
|
| nsv527866 | Chr.16:4206462 - 4412618 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map