Assay Details
Target Gene Details
Entrez Gene ID: | 200373 |
Gene Name: | cilia and flagella associated protein 221 |
Gene Aliases: |
FAP221, PCDP1 |
Location: |
Chr.2:119544392-119688610 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CFAP221 | NM_001271049.1 | NP_001257978.1 | ||
| NR_073132.1 | ||||
| NR_073133.1 | ||||
| XM_006712353.3 | XP_006712416.1 | |||
| XM_017003550.1 | XP_016859039.1 | |||
| XM_017003551.1 | XP_016859040.1 | |||
| XM_017003552.1 | XP_016859041.1 | |||
| XM_017003553.1 | XP_016859042.1 | |||
| XM_017003554.1 | XP_016859043.1 | |||
| XM_017003555.1 | XP_016859044.1 | |||
| XM_017003556.1 | XP_016859045.1 | |||
| XM_017003557.1 | XP_016859046.1 | |||
| XM_017003558.1 | XP_016859047.1 | |||
| AK293665.1 | ||||
| AK304416.1 | ||||
| BC036530.1 | AAH36530.1 | |||
| DC417687.1 |
More Information
Set Membership: |
|

Genomic Map