Assay Details
Target Gene Details
Entrez Gene ID: | 79573 |
Gene Name: | tetratricopeptide repeat domain 13 |
Gene Aliases: |
- |
Location: |
Chr.1:230906241-230978872 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 22 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3589093 | Chr.1:230482037 - 231277190 on Build GRCh38 | Gain |
|
| nsv1000488 | Chr.1:230799658 - 231269735 on Build GRCh38 | Gain |
|

Genomic Map