Assay Details
Target Gene Details
Entrez Gene ID: | 11145 |
Gene Name: | phospholipase A2 group XVI |
Gene Aliases: |
AdPLA, H-REV107-1, HRASLS3, HREV107, HREV107-1, HREV107-3, HRSL3 |
Location: |
Chr.11:63574462-63616926 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLA2G16 | NM_001128203.1 | 4 | 482 | NP_001121675.1 |
NM_007069.3 | 3 | 766 | NP_009000.2 | |
XM_006718426.1 | 4 | 684 | XP_006718489.1 | |
XM_011544741.1 | 3 | 668 | XP_011543043.1 | |
AB030814.1 | 3 | 392 | BAB08108.1 | |
AB439591.1 | 2 | 343 | ||
AF317086.1 | 3 | 397 | AAL26892.1 | |
AI590735.1 | 2 | 320 | ||
AK313075.1 | 3 | 765 | ||
AX775893.1 | 3 | 397 | CAE11646.1 | |
BC001387.2 | 3 | 698 | AAH01387.1 | |
BC103807.2 | 3 | 487 | AAI03808.1 | |
BC103808.2 | 3 | 437 | AAI03809.1 | |
BG719903.1 | 4 | 477 | ||
CK003948.1 | 4 | 483 | ||
X92814.1 | 3 | 750 | CAA63423.1 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
