Assay Details
Target Gene Details
Entrez Gene ID: | 81887 |
Gene Name: | LAS1 like, ribosome biogenesis factor |
Gene Aliases: |
Las1-like, dJ475B7.2 |
Location: |
Chr.X:65512582-65534806 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LAS1L | NM_001170649.1 | 2 | 398 | NP_001164120.1 |
| NM_001170650.1 | NP_001164121.1 | |||
| NM_031206.4 | 2 | 398 | NP_112483.1 | |
| XM_005262301.1 | 2 | 383 | XP_005262358.1 | |
| XM_005262304.1 | 2 | 383 | XP_005262361.1 | |
| XM_005262305.3 | 2 | 383 | XP_005262362.1 | |
| XM_005262306.3 | 2 | 383 | XP_005262363.1 | |
| XM_011531045.1 | XP_011529347.1 | |||
| XM_011531046.1 | 2 | 383 | XP_011529348.1 | |
| XM_017029877.1 | 2 | 383 | XP_016885366.1 | |
| XM_017029878.1 | 2 | 383 | XP_016885367.1 | |
| XM_017029879.1 | 2 | 383 | XP_016885368.1 | |
| XM_017029880.1 | 2 | 383 | XP_016885369.1 | |
| AF116730.1 | 2 | 336 | AAO15306.1 | |
| AK022587.1 | 2 | 348 | BAB14114.1 | |
| AK074087.1 | 2 | 367 | BAB84913.1 | |
| AK097383.1 | ||||
| AK225059.1 | 2 | 331 | ||
| AK295591.1 | 2 | 348 | ||
| BC014545.1 | 2 | 382 | AAH14545.1 | |
| BC018610.1 | 2 | 346 | AAH18610.1 | |
| BC019302.1 | 2 | 343 | AAH19302.1 | |
| DA619280.1 | 2 | 399 |
More Information
Set Membership: |
|

Genomic Map