Assay Details
Target Gene Details
Entrez Gene ID: | 1233 |
Gene Name: | C-C motif chemokine receptor 4 |
Gene Aliases: |
CC-CKR-4, CD194, CKR4, CMKBR4, ChemR13, HGCN:14099, K5-5 |
Location: |
Chr.3:32951555-32955312 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCR4 | NM_005508.4 | 2 | 206 | NP_005499.1 |
| XM_017005687.1 | 2 | 253 | XP_016861176.1 | |
| AK292674.1 | 2 | 136 | ||
| BC069139.1 | 1 | 91 | AAH69139.1 | |
| BC071751.1 | 2 | 206 | AAH71751.1 | |
| BC074935.2 | 1 | 66 | AAH74935.1 | |
| CD639869.1 | 2 | 235 | ||
| CD640816.1 | 2 | 209 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1003285 | Chr.3:31386933 - 33172886 on Build GRCh38 | Gain |
|
| nsv998302 | Chr.3:32595752 - 33112754 on Build GRCh38 | Gain |
|
| nsv508909 | Chr.3:32927304 - 32956808 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map