Assay Details
Target Gene Details
Entrez Gene ID: | 771 |
Gene Name: | carbonic anhydrase 12 |
Gene Aliases: |
CA-XII, CAXII, HsT18816, T18816 |
Location: |
Chr.15:63321378-63382166 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CA12 | NM_001218.4 | 11 | 3596 | NP_001209.1 |
| NM_001293642.1 | 9 | 3383 | NP_001280571.1 | |
| NM_206925.2 | 10 | 3563 | NP_996808.1 | |
| NR_135511.1 | 10 | 3666 | ||
| AK000158.1 | 1 | 312 | BAA90981.1 | |
| BC001012.2 | 1 | 453 | AAH01012.1 | |
| BM994285.1 | 1 | 345 | ||
| CA309380.1 | 1 | 631 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1035441 | Chr.15:62406037 - 63349997 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map