Assay Details
Target Gene Details
Entrez Gene ID: | 56477 |
Gene Name: | C-C motif chemokine ligand 28 |
Gene Aliases: |
CCK1, MEC, SCYA28 |
Location: |
Chr.5:43356972-43412391 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCL28 | NM_001301873.1 | 2 | 220 | NP_001288802.1 |
| NM_001301874.1 | 2 | 220 | NP_001288803.1 | |
| NM_001301875.1 | 2 | 311 | NP_001288804.1 | |
| NM_148672.3 | 2 | 220 | NP_683513.1 | |
| AF110384.1 | 2 | 199 | AAG43193.1 | |
| AF220210.1 | 2 | 145 | AAF87205.1 | |
| AF266504.1 | 2 | 197 | AAG16691.1 | |
| AK307307.1 | 2 | 175 | ||
| AK312120.1 | 2 | 203 | ||
| BC062668.1 | 2 | 162 | AAH62668.1 | |
| BC069532.1 | 2 | 195 | AAH69532.1 | |
| BX486632.1 | 2 | 221 | ||
| CD671712.1 | 2 | 311 | ||
| HM067700.1 | 2 | 145 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508352 | Chr.5:43237728 - 43390960 on Build GRCh38 | Deletion |
|
| esv3604897 | Chr.5:43343819 - 43405322 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map