Assay Details
Target Gene Details
Entrez Gene ID: | 8404 |
Gene Name: | SPARC like 1 |
Gene Aliases: |
MAST 9, MAST9, PIG33, SC1 |
Location: |
Chr.4:87473330-87529503 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPARCL1 | NM_001128310.2 | 5 | 1278 | NP_001121782.1 |
| NM_001291976.1 | 5 | 1285 | NP_001278905.1 | |
| NM_001291977.1 | 2 | 933 | NP_001278906.1 | |
| NM_004684.5 | 4 | 1176 | NP_004675.3 | |
| AK294335.1 | 5 | 633 | ||
| AK298039.1 | ||||
| AK300956.1 | 5 | 585 | ||
| AK304494.1 | 2 | 802 | ||
| AK304628.1 | ||||
| AK316533.1 | 5 | 875 | ||
| AL833430.1 | 4 | 914 | ||
| AY513278.1 | 4 | 786 | AAT08031.1 | |
| BC033721.1 | 4 | 951 | AAH33721.1 | |
| BX647713.1 | 5 | 827 | ||
| X82157.1 | 4 | 903 | CAA57650.1 | |
| X86693.1 | 4 | 1028 | CAA60386.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3601276 | Chr.4:87422870 - 87515781 on Build GRCh38 | Loss |
|
| esv3601278 | Chr.4:87455460 - 87496054 on Build GRCh38 | Loss |
|
| nsv1002789 | Chr.4:87318188 - 87685333 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map