Assay Details
Target Gene Details
Entrez Gene ID: | 81608 |
Gene Name: | factor interacting with PAPOLA and CPSF1 |
Gene Aliases: |
FIP1, Rhe, hFip1 |
Location: |
Chr.4:53377572-53462611 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FIP1L1 | NM_001134937.1 | NP_001128409.1 | ||
| NM_001134938.1 | NP_001128410.1 | |||
| NM_030917.3 | NP_112179.2 | |||
| XM_005265769.4 | 19 | 2502 | XP_005265826.1 | |
| XM_005265773.4 | 18 | 2435 | XP_005265830.1 | |
| XM_005265774.4 | 17 | 2347 | XP_005265831.1 | |
| XM_005265778.4 | 18 | 2273 | XP_005265835.1 | |
| XM_005265779.4 | 17 | 2201 | XP_005265836.1 | |
| XM_005265781.4 | 17 | 2319 | XP_005265838.1 | |
| XM_005265782.4 | 16 | 2238 | XP_005265839.1 | |
| XM_017008662.1 | 17 | 2380 | XP_016864151.1 | |
| XM_017008663.1 | 18 | 2336 | XP_016864152.1 | |
| XM_017008664.1 | 17 | 2391 | XP_016864153.1 | |
| XM_017008665.1 | 17 | 2307 | XP_016864154.1 | |
| XM_017008666.1 | 16 | 2324 | XP_016864155.1 | |
| XM_017008667.1 | 17 | 2290 | XP_016864156.1 | |
| XM_017008668.1 | 16 | 2336 | XP_016864157.1 | |
| XM_017008669.1 | 16 | 2236 | XP_016864158.1 | |
| XM_017008670.1 | 16 | 2132 | XP_016864159.1 | |
| XM_017008671.1 | 16 | 2051 | XP_016864160.1 | |
| AK090938.1 | 7 | 1202 |
Target Gene Details
Entrez Gene ID: | 84708 |
Gene Name: | ligand of numb-protein X 1 |
Gene Aliases: |
LNX, MPDZ, PDZRN2 |
Location: |
Chr.4:53458264-53652524 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LNX1 | XM_005265785.4 | 11 | 3651 | XP_005265842.1 |
| XM_017008776.1 | 12 | 3712 | XP_016864265.1 | |
| AF237782.1 | 10 | 3155 | AAK15039.1 |
More Information
Set Membership: |
|

Genomic Map