Assay Details
Target Gene Details
Entrez Gene ID: | 3690 |
Gene Name: | integrin subunit beta 3 |
Gene Aliases: |
BDPLT16, BDPLT2, CD61, GP3A, GPIIIa, GT |
Location: |
Chr.17:47253842-47312711 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITGB3 | NM_000212.2 | 15 | 3364 | NP_000203.2 |
| BE389074.1 | 2 | 384 | ||
| M20311.1 | 15 | 3360 | AAA60122.1 | |
| M35999.1 | 15 | 3362 | AAA35927.1 | |
| U95204.1 | AAB71380.1 |
Target Gene Details
Entrez Gene ID: | 102724508 |
Gene Name: | thyroid cancer-associated transcript 158 |
Gene Aliases: |
- |
Location: |
Chr.17:47303443-47323901 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THCAT158 | NR_110880.1 | |||
| NR_110881.1 | ||||
| AA430297.1 | ||||
| AW293117.1 | ||||
| HY036509.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs73322319] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map