Assay Details
Target Gene Details
Entrez Gene ID: | 3783 |
Gene Name: | potassium calcium-activated channel subfamily N member 4 |
Gene Aliases: |
DHS2, IK, IK1, IKCA1, KCA4, KCa3.1, SK4, hIKCa1, hKCa4, hSK4 |
Location: |
Chr.19:43766533-43782139 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNN4 | NM_002250.2 | 9 | 2049 | NP_002241.1 |
| XM_005258882.2 | 8 | 2835 | XP_005258939.1 | |
| XM_005258883.2 | 9 | 2927 | XP_005258940.1 | |
| AF022797.1 | 9 | 2049 | AAC51913.1 | |
| AF033021.1 | 9 | 1777 | AAC36804.1 | |
| BC015337.1 | 9 | 1662 | AAH15337.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509745 | Chr.19:43746288 - 43808495 on Build GRCh38 | Insertion |
|
| nsv833839 | Chr.19:43595661 - 43768147 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map