Assay Details
Target Gene Details
Entrez Gene ID: | 54093 |
Gene Name: | SET domain containing 4 |
Gene Aliases: |
C21orf18, C21orf27 |
Location: |
Chr.21:36034541-36060818 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SETD4 | NM_001286752.1 | NP_001273681.1 | ||
| NM_017438.4 | NP_059134.1 | |||
| NR_040087.2 | ||||
| XM_011529636.1 | XP_011527938.1 | |||
| XM_011529637.1 | XP_011527939.1 | |||
| XM_011529638.2 | XP_011527940.1 | |||
| XM_011529639.1 | XP_011527941.1 | |||
| XM_011529640.2 | XP_011527942.1 | |||
| XM_011529642.1 | XP_011527944.1 | |||
| XM_011529643.1 | XP_011527945.1 | |||
| XM_011529644.1 | XP_011527946.1 | |||
| XM_017028403.1 | XP_016883892.1 | |||
| XM_017028404.1 | XP_016883893.1 | |||
| AF391112.1 | AAL34503.1 | |||
| AI636385.1 | ||||
| AK001660.1 | BAA91819.1 | |||
| AK095215.1 | ||||
| AK124254.1 | ||||
| AK126368.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470899 | Chr.21:36001424 - 36096978 on Build GRCh38 | Loss |
|
| nsv459247 | Chr.21:36003345 - 36087866 on Build GRCh38 | Loss |
|
| nsv3504 | Chr.21:36007961 - 36040889 on Build GRCh38 | Insertion |
|
| esv3646959 | Chr.21:36032869 - 36035021 on Build GRCh38 | Loss |
|
| nsv964547 | Chr.21:36003423 - 36090261 on Build GRCh38 | Deletion |
|
| esv2665899 | Chr.21:36032796 - 36035080 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map