Assay Details
Target Gene Details
Entrez Gene ID: | 5309 |
Gene Name: | paired like homeodomain 3 |
Gene Aliases: |
ASMD, ASOD, CTPP4, CTRCT11, PTX3 |
Location: |
Chr.10:102230189-102241474 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PITX3 | NM_005029.3 | 4 | 662 | NP_005020.1 |
| AF041339.1 | 3 | 511 | AAC24502.1 | |
| BC011642.2 | 4 | 661 | AAH11642.1 |
More Information
Set Membership: |
|

Genomic Map