Assay Details
Target Gene Details
Entrez Gene ID: | 4914 |
Gene Name: | neurotrophic receptor tyrosine kinase 1 |
Gene Aliases: |
MTC, TRK, TRK1, TRKA, Trk-A, p140-TrkA |
Location: |
Chr.1:156815750-156881850 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NTRK1 | NM_001007792.1 | NP_001007793.1 | ||
| AK126428.1 |
Target Gene Details
Entrez Gene ID: | 9047 |
Gene Name: | SH2 domain containing 2A |
Gene Aliases: |
F2771, SCAP, TSAD, VRAP |
Location: |
Chr.1:156806238-156816853 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SH2D2A | NM_001161441.1 | NP_001154913.1 | ||
| NM_001161442.1 | NP_001154914.1 | |||
| NM_001161443.1 | 1 | 80 | NP_001154915.1 | |
| NM_001161444.1 | NP_001154916.1 | |||
| NM_003975.3 | NP_003966.2 | |||
| XM_006711615.2 | 2 | 219 | XP_006711678.1 | |
| XM_017002762.1 | 2 | 218 | XP_016858251.1 | |
| XM_017002763.1 | 2 | 221 | XP_016858252.1 | |
| XM_017002764.1 | XP_016858253.1 | |||
| XM_017002765.1 | XP_016858254.1 | |||
| XM_017002766.1 | XP_016858255.1 | |||
| XM_017002767.1 | 2 | 224 | XP_016858256.1 | |
| AF051325.1 | AAC99298.1 | |||
| AF097744.1 | AAF43260.1 | |||
| AJ000553.1 | CAA04185.1 | |||
| AK222737.1 | BAD96457.1 | |||
| AK313706.1 | ||||
| AY763098.1 | 1 | 80 | AAV34673.1 | |
| AY763099.1 | AAV34674.1 | |||
| AY763100.1 | AAV34675.1 | |||
| BC012107.1 | AAH12107.1 | |||
| DB128222.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831648 | Chr.1:156694085 - 156912126 on Build GRCh38 | Loss |
|
| nsv831659 | Chr.1:156771159 - 156980204 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map