Assay Details
Target Gene Details
Entrez Gene ID: | 83591 |
Gene Name: | THAP domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.12:71663897-71680648 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THAP2 | NM_031435.3 | NP_113623.1 | ||
| BP371303.1 |
Target Gene Details
Entrez Gene ID: | 196441 |
Gene Name: | zinc finger C3H1-type containing |
Gene Aliases: |
CCDC131, CSRC2, PSRC2 |
Location: |
Chr.12:71609599-71663969 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZFC3H1 | NM_144982.4 | 1 | 99 | NP_659419.3 |
| AB011118.2 | BAA25472.2 | |||
| AI015547.1 | 1 | 99 | ||
| BC064336.1 | AAH64336.1 | |||
| BC073843.1 | AAH73843.1 | |||
| BX647709.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832451 | Chr.12:71584552 - 71806081 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map