Assay Details
Target Gene Details
Entrez Gene ID: | 25963 |
Gene Name: | transmembrane protein 87A |
Gene Aliases: |
- |
Location: |
Chr.15:42210452-42273584 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM87A | NM_001286487.1 | 19 | 2187 | NP_001273416.1 |
| NM_015497.4 | 20 | 2248 | NP_056312.2 | |
| XM_005254287.1 | 20 | 2102 | XP_005254344.1 | |
| XM_006720468.2 | 20 | 2107 | XP_006720531.1 | |
| XM_011521443.2 | 19 | 2019 | XP_011519745.1 | |
| XM_017022067.1 | 20 | 2104 | XP_016877556.1 | |
| AF129927.1 | 4 | 584 | ||
| AF132733.1 | 20 | 2104 | AAF66444.1 | |
| AI023965.1 | ||||
| AK074870.1 | 20 | 2097 | BAC11256.1 | |
| AK075403.1 | 20 | 2134 | BAC11598.1 | |
| AK290484.1 | ||||
| AK297816.1 | ||||
| AL049944.1 | 7 | 794 | CAB43218.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1054102 | Chr.15:42200854 - 42236567 on Build GRCh38 | Gain |
|
| nsv832985 | Chr.15:42079284 - 42272768 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map