Assay Details
Target Gene Details
Entrez Gene ID: | 151516 |
Gene Name: | aspartic peptidase, retroviral-like 1 |
Gene Aliases: |
MUNO, SASP, SASPase, Taps |
Location: |
Chr.2:69960089-69962265 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASPRV1 | NM_152792.2 | 1 | 798 | NP_690005.2 |
| AK055994.1 | 1 | 798 | BAB71067.1 | |
| AK057813.1 | 1 | 198 | BAB71587.1 | |
| BC094000.1 | 1 | 316 | AAH94000.1 | |
| BC094002.1 | 1 | 316 | AAH94002.1 |
More Information
Set Membership: |
|

Genomic Map