Assay Details
Target Gene Details
Entrez Gene ID: | 1030 |
Gene Name: | cyclin dependent kinase inhibitor 2B |
Gene Aliases: |
CDK4I, INK4B, MTS2, P15, TP15, p15INK4b |
Location: |
Chr.9:22002903-22009313 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDKN2B | NM_004936.3 | NP_004927.2 | ||
| NM_078487.2 | NP_511042.1 | |||
| AF004819.1 | AAB69989.1 | |||
| BC014469.1 | AAH14469.1 | |||
| BI560960.1 | ||||
| CR536529.1 | CAG38766.1 | |||
| JQ694046.1 | ||||
| JQ694047.1 | ||||
| JQ694048.1 | ||||
| JX391994.1 | ||||
| L36844.1 | AAA50282.1 | |||
| U17075.1 | AAC50075.1 |
Target Gene Details
Entrez Gene ID: | 100048912 |
Gene Name: | CDKN2B antisense RNA 1 |
Gene Aliases: |
ANRIL, CDKN2B-AS, CDKN2BAS, NCRNA00089, PCAT12, p15AS |
Location: |
Chr.9:21994791-22121097 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDKN2B-AS1 | NR_003529.3 | |||
| NR_047532.1 | ||||
| NR_047533.1 | ||||
| NR_047534.1 | ||||
| NR_047535.1 | ||||
| NR_047536.1 | ||||
| NR_047537.1 | ||||
| NR_047538.1 | ||||
| NR_047539.1 | ||||
| NR_047540.1 | ||||
| NR_047541.1 | ||||
| NR_047542.1 | ||||
| NR_047543.1 | ||||
| NR_120536.1 | ||||
| AB548314.1 | ||||
| BI860238.1 | ||||
| CB109081.1 | ||||
| DQ485453.1 | ||||
| DQ485454.1 | ||||
| EU741058.1 | ||||
| GQ422823.1 | ||||
| GQ495918.1 | ||||
| GQ495919.1 | ||||
| GQ495920.1 | ||||
| GQ495921.1 | ||||
| GQ495922.1 | ||||
| GQ495923.1 | ||||
| GQ495924.1 | ||||
| GQ495925.1 | ||||
| GQ495926.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1018514 | Chr.9:21944276 - 22075327 on Build GRCh38 | Gain |
|
| nsv613760 | Chr.9:22006270 - 22009961 on Build GRCh38 | Loss |
|
| esv2764150 | Chr.9:20659506 - 22349184 on Build GRCh38 | Gain+Loss |
|
| nsv8426 | Chr.9:21763811 - 22176201 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map