Assay Details
Target Gene Details
Entrez Gene ID: | 80208 |
Gene Name: | spastic paraplegia 11 (autosomal recessive) |
Gene Aliases: |
ALS5, CMT2X, KIAA1840 |
Location: |
Chr.15:44562696-44663678 on Build GRCh38 |
Assay Gene Location: | Within Exon 41 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPG11 | NM_001160227.1 | 38 | 7122 | NP_001153699.1 |
| NM_025137.3 | 40 | 7461 | NP_079413.3 | |
| XM_006720700.1 | 40 | 7317 | XP_006720763.1 | |
| XM_017022634.1 | 39 | 7353 | XP_016878123.1 | |
| XM_017022636.1 | 29 | 5270 | XP_016878125.1 | |
| AB058743.2 | 40 | 7431 | BAB47469.2 | |
| AK025092.1 | 11 | 1582 | BAB15065.1 | |
| AK057869.1 | 8 | 1801 | ||
| AK225186.1 | 11 | 1582 | ||
| BC024161.1 | 10 | 1547 | ||
| BC067798.1 | 11 | 1823 | AAH67798.1 | |
| BC094704.1 | 11 | 1955 | AAH94704.1 | |
| BC150640.1 | 38 | 7119 | ||
| BC153879.1 | 40 | 7431 |
More Information
Set Membership: |
|

Genomic Map