Assay Details
Target Gene Details
Entrez Gene ID: | 51280 |
Gene Name: | golgi membrane protein 1 |
Gene Aliases: |
C9orf155, GOLPH2, GP73, HEL46, PSEC0257, bA379P1.3 |
Location: |
Chr.9:86026141-86100201 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GOLM1 | NM_016548.3 | 10 | 1437 | NP_057632.2 |
| NM_177937.2 | 10 | 1429 | NP_808800.1 | |
| AF236056.1 | 10 | 1378 | AAF44663.1 | |
| AK027717.1 | 10 | 1400 | ||
| AK075542.1 | 10 | 1371 | BAC11685.1 | |
| AK075557.1 | 10 | 1341 | ||
| AK092943.1 | 11 | 1317 | ||
| AY358593.1 | 10 | 1342 | AAQ88956.1 | |
| BC001740.1 | 10 | 1352 | AAH01740.1 | |
| CR457201.1 | CAG33482.1 | |||
| EU794616.1 | 10 | 1396 |
More Information
Set Membership: |
|

Genomic Map