Assay Details
Target Gene Details
Entrez Gene ID: | 10693 |
Gene Name: | chaperonin containing TCP1 subunit 6B |
Gene Aliases: |
CCT-zeta-2, CCTZ-2, Cctz2, TCP-1-zeta-2, TSA303 |
Location: |
Chr.17:34927859-34981221 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCT6B | XM_011524203.2 | XP_011522505.1 | ||
| XM_017024023.1 | XP_016879512.1 | |||
| XM_017024024.1 | XP_016879513.1 |
Target Gene Details
Entrez Gene ID: | 91603 |
Gene Name: | zinc finger protein 830 |
Gene Aliases: |
CCDC16, OMCG1 |
Location: |
Chr.17:34961530-34963186 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF830 | NM_052857.3 | 1 | 885 | NP_443089.3 |
| AK055707.1 | 1 | 869 | BAB70992.1 | |
| AK315671.1 | 1 | 885 | ||
| BC002913.2 | 1 | 830 | AAH02913.2 | |
| BC009044.1 | 1 | 448 | AAH09044.1 | |
| BC011584.1 | 1 | 830 | ||
| BM720333.1 |
More Information
Set Membership: |
|

Genomic Map