Assay Details
Target Gene Details
Entrez Gene ID: | 5051 |
Gene Name: | platelet activating factor acetylhydrolase 2 |
Gene Aliases: |
HSD-PLA2 |
Location: |
Chr.1:25959764-25998164 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAFAH2 | NM_000437.3 | 7 | 783 | NP_000428.2 |
| XM_006710669.3 | 4 | 1075 | XP_006710732.1 | |
| XM_006710670.3 | 7 | 848 | XP_006710733.1 | |
| XM_011541528.2 | 8 | 914 | XP_011539830.1 | |
| XM_011541529.2 | 5 | 536 | XP_011539831.1 | |
| XM_017001390.1 | 7 | 809 | XP_016856879.1 | |
| XM_017001391.1 | 6 | 584 | XP_016856880.1 | |
| XM_017001392.1 | 6 | 655 | XP_016856881.1 | |
| AK313815.1 | 7 | 710 | ||
| BC001158.1 | 7 | 728 | AAH01158.1 | |
| D87845.1 | 7 | 806 | BAA13468.1 | |
| U89386.1 | 7 | 682 | AAC39707.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834502 | Chr.1:25834281 - 25987428 on Build GRCh38 | Gain |
|
| nsv834513 | Chr.1:25922980 - 26087385 on Build GRCh38 | Gain |
|
| nsv1160537 | Chr.1:25970542 - 25992341 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map