Assay Details
Target Gene Details
Entrez Gene ID: | 10911 |
Gene Name: | urotensin 2 |
Gene Aliases: |
PRO1068, U-II, UCN2, UII |
Location: |
Chr.1:7846345-7913615 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UTS2 | NM_006786.3 | NP_006777.1 | ||
| NM_021995.2 | NP_068835.1 | |||
| XM_011540537.2 | XP_011538839.1 | |||
| XM_011540538.1 | XP_011538840.1 | |||
| XM_017000116.1 | XP_016855605.1 | |||
| XM_017000117.1 | XP_016855606.1 | |||
| XM_017000118.1 | XP_016855607.1 | |||
| XM_017000119.1 | XP_016855608.1 | |||
| AF104118.1 | AAD13070.1 | |||
| AY358375.1 | AAQ88741.1 | |||
| BC126443.1 |
More Information
Set Membership: |
|

Genomic Map