Assay Details
Target Gene Details
Entrez Gene ID: | 27 |
Gene Name: | ABL proto-oncogene 2, non-receptor tyrosine kinase |
Gene Aliases: |
ABLL, ARG |
Location: |
Chr.1:179099327-179229693 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABL2 | NM_001136000.2 | 13 | 3112 | NP_001129472.1 |
| NM_001168236.1 | 11 | 3645 | NP_001161708.1 | |
| NM_001168237.1 | 13 | 3399 | NP_001161709.1 | |
| NM_001168238.1 | 12 | 3336 | NP_001161710.1 | |
| NM_001168239.1 | 12 | 3049 | NP_001161711.1 | |
| NM_005158.4 | 12 | 3421 | NP_005149.4 | |
| NM_007314.3 | 12 | 3708 | NP_009298.1 | |
| XM_005245088.2 | 11 | 3343 | XP_005245145.1 | |
| XM_017001035.1 | 11 | 6018 | XP_016856524.1 | |
| BC065912.1 | 12 | 3388 | AAH65912.1 | |
| BX538317.1 | 13 | 3151 | CAD98092.1 | |
| FJ542283.1 | 12 | 3004 | ||
| FJ542284.1 | 13 | 3112 | ||
| FJ542285.1 | 11 | 3358 | ||
| FJ542286.1 | 12 | 3049 | ||
| M35296.1 | 12 | 3625 | AAA35553.1 |
More Information
Set Membership: |
|

Genomic Map