Assay Details
Target Gene Details
Entrez Gene ID: | 55761 |
Gene Name: | tetratricopeptide repeat domain 17 |
Gene Aliases: |
- |
Location: |
Chr.11:43358885-43494933 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC17 | NM_001307943.1 | NP_001294872.1 | ||
| NM_018259.5 | NP_060729.2 | |||
| XM_005253016.1 | XP_005253073.1 | |||
| XM_011520218.2 | XP_011518520.1 | |||
| XM_017018010.1 | XP_016873499.1 | |||
| AK001752.1 | BAA91883.1 | |||
| BC017238.2 | AAH17238.1 | |||
| BC033000.1 | AAH33000.1 | |||
| DA106610.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv554119 | Chr.11:42530505 - 43398360 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map