Assay Details
Target Gene Details
Entrez Gene ID: | 85027 |
Gene Name: | small integral membrane protein 3 |
Gene Aliases: |
C5orf62, MST150, NID67 |
Location: |
Chr.5:150777946-150796736 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMIM3 | NM_032947.4 | NP_116565.3 | ||
| AF313413.1 | AAG53959.1 | |||
| AK027847.1 | ||||
| AK125286.1 | BAC86114.1 | |||
| BC009719.1 | AAH09719.1 | |||
| BC106040.1 | AAI06041.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759386 | Chr.5:150445030 - 150906180 on Build GRCh38 | Loss |
|
| nsv428129 | Chr.5:150641519 - 150906180 on Build GRCh38 | Loss |
|
| nsv5067 | Chr.5:150785595 - 150853767 on Build GRCh38 | Deletion |
|
| nsv435820 | Chr.5:150795893 - 150802712 on Build GRCh38 | Deletion |
|
| dgv5817n100 | Chr.5:150777463 - 150933758 on Build GRCh38 | Loss |
|
| dgv5818n100 | Chr.5:150789498 - 150844521 on Build GRCh38 | Loss |
|
| dgv10114n54 | Chr.5:150777798 - 150937675 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map