Assay Details
Target Gene Details
Entrez Gene ID: | 5790 |
Gene Name: | protein tyrosine phosphatase, receptor type C associated protein |
Gene Aliases: |
CD45-AP, LPAP |
Location: |
Chr.11:67435510-67437682 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTPRCAP | NM_005608.2 | NP_005599.1 | ||
| AK312022.1 | ||||
| BC113859.1 | AAI13860.1 | |||
| BC114467.1 | AAI14468.1 | |||
| X81422.1 | CAA57182.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524096 | Chr.11:67418529 - 67457517 on Build GRCh38 | Loss |
|
| dgv1977n54 | Chr.11:67397024 - 67521123 on Build GRCh38 | Loss |
|
| dgv52n111 | Chr.11:67362651 - 67521123 on Build GRCh38 | Deletion |
|
| nsv521723 | Chr.11:67408221 - 67543787 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map