Assay Details
Target Gene Details
Entrez Gene ID: | 80347 |
Gene Name: | Coenzyme A synthase |
Gene Aliases: |
DPCK, NBIA6, NBP, PPAT, UKR1, pOV-2 |
Location: |
Chr.17:42562074-42566281 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COASY | NM_001042529.2 | 2 | 238 | NP_001035994.1 |
| NM_001042532.3 | 3 | 370 | NP_001035997.2 | |
| NM_025233.6 | 1 | 630 | NP_079509.5 | |
| XM_006722116.3 | 2 | 264 | XP_006722179.1 | |
| XM_011525300.1 | 2 | 434 | XP_011523602.1 | |
| XM_017025167.1 | 1 | 574 | XP_016880656.1 | |
| XM_017025168.1 | XP_016880657.1 | |||
| AF453478.1 | 1 | 81 | AAL50813.1 | |
| AK075415.1 | 1 | 537 | ||
| AK225485.1 | 2 | 167 | ||
| AK297153.1 | 3 | 293 | ||
| AK303766.1 | 2 | 161 | ||
| AK314076.1 | 3 | 294 | ||
| AL360168.1 | 2 | 412 | ||
| AL583142.3 | 2 | 156 | ||
| AL833588.1 | 2 | 162 | ||
| AY094602.1 | 2 | 191 | AAM19996.1 | |
| BC067254.1 | 1 | 534 | AAH67254.1 | |
| BM473635.1 | 2 | 411 | ||
| BM781531.1 | 2 | 417 | ||
| DA131866.1 | ||||
| DA786254.1 | 2 | 238 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064012 | Chr.17:42427786 - 42606703 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map