Assay Details
Target Gene Details
Entrez Gene ID: | 377047 |
Gene Name: | protease, serine 45 |
Gene Aliases: |
TESSP5 |
Location: |
Chr.3:46742091-46744755 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRSS45 | NM_199183.2 | NP_954652.2 | ||
| BC133695.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516623 | Chr.3:46638573 - 46811189 on Build GRCh38 | Loss |
|
| esv33334 | Chr.3:46736319 - 46749941 on Build GRCh38 | Gain |
|
| nsv590179 | Chr.3:46687169 - 46748907 on Build GRCh38 | Loss |
|
| esv2759147 | Chr.3:46638962 - 46930999 on Build GRCh38 | Loss |
|
| dgv4730n100 | Chr.3:46729944 - 46808087 on Build GRCh38 | Loss |
|
| dgv4731n100 | Chr.3:46733893 - 46827326 on Build GRCh38 | Loss |
|
| nsv3798 | Chr.3:46737140 - 46824116 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map