Assay Details
Target Gene Details
Entrez Gene ID: | 57489 |
Gene Name: | outer dense fiber of sperm tails 2 like |
Gene Aliases: |
dJ977L11.1 |
Location: |
Chr.1:86346824-86396345 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ODF2L | NM_001007022.2 | 8 | 1120 | NP_001007023.2 |
| NM_001184765.1 | 8 | 1127 | NP_001171694.1 | |
| NM_001184766.1 | 8 | 1127 | NP_001171695.1 | |
| NM_020729.2 | 8 | 1127 | NP_065780.2 | |
| XM_005271054.3 | 8 | 1125 | XP_005271111.1 | |
| XM_011541815.2 | 8 | 1125 | XP_011540117.1 | |
| XM_011541819.2 | 8 | 1125 | XP_011540121.1 | |
| XM_011541821.2 | 8 | 1125 | XP_011540123.1 | |
| XM_011541823.2 | 8 | 1125 | XP_011540125.1 | |
| XM_011541824.2 | 8 | 1125 | XP_011540126.1 | |
| XM_017001876.1 | 8 | 917 | XP_016857365.1 | |
| XM_017001877.1 | 8 | 917 | XP_016857366.1 | |
| XM_017001878.1 | 7 | 1008 | XP_016857367.1 | |
| XM_017001879.1 | 8 | 1125 | XP_016857368.1 | |
| XM_017001880.1 | 7 | 1008 | XP_016857369.1 | |
| XM_017001881.1 | 6 | 880 | XP_016857370.1 | |
| AB033055.1 | 8 | 1040 | ||
| AK302792.1 | 8 | 1099 | ||
| AK303209.1 | 8 | 1124 | ||
| AK308351.1 | 5 | 650 | ||
| BC009779.1 | 4 | 524 | AAH09779.1 | |
| BC029420.1 | 8 | 845 | ||
| BC047368.1 | 8 | 1067 | AAH47368.1 | |
| BC091490.1 | 8 | 1026 | AAH91490.1 | |
| BX499763.1 | 4 | 341 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv546722 | Chr.1:85812632 - 86425390 on Build GRCh38 | Loss |
|
| nsv1002541 | Chr.1:86223405 - 86440219 on Build GRCh38 | Gain |
|

Genomic Map