Assay Details
Target Gene Details
Entrez Gene ID: | 339983 |
Gene Name: | N-acetyltransferase 8 like |
Gene Aliases: |
CML3, NACED, NAT8-LIKE |
Location: |
Chr.4:2059512-2069089 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAT8L | NM_178557.3 | NP_848652.2 | ||
| AK094797.1 | BAC04426.1 | |||
| BC093906.1 | AAH93906.1 | |||
| BC093908.1 | AAH93908.1 | |||
| BC103748.1 | AAI03749.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2677287 | Chr.4:2057718 - 2061365 on Build GRCh38 | Deletion |
|
| esv2726835 | Chr.4:2019561 - 2959568 on Build GRCh38 | Deletion |
|
| nsv1145019 | Chr.4:2058673 - 2061173 on Build GRCh38 | Deletion |
|
| dgv8909n54 | Chr.4:2058874 - 2075739 on Build GRCh38 | Loss |
|
| nsv949757 | Chr.4:2056974 - 2082173 on Build GRCh38 | Deletion |
|
| nsv822441 | Chr.4:1991240 - 2080440 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map