Assay Details
Target Gene Details
Entrez Gene ID: | 9644 |
Gene Name: | SH3 and PX domains 2A |
Gene Aliases: |
FISH, SH3MD1, TKS5 |
Location: |
Chr.10:103594027-103855464 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SH3PXD2A | NM_014631.2 | 14 | 8471 | NP_055446.2 |
| XM_005270294.4 | 15 | 8604 | XP_005270351.1 | |
| XM_005270295.4 | 14 | 8558 | XP_005270352.1 | |
| XM_005270297.3 | 9 | 9835 | XP_005270354.1 | |
| XM_005270298.4 | 9 | 8638 | XP_005270355.1 | |
| XM_006718079.3 | 16 | 8701 | XP_006718142.1 | |
| XM_006718080.3 | 10 | 8732 | XP_006718143.1 | |
| XM_011540385.2 | 15 | 8655 | XP_011538687.1 | |
| XM_011540386.2 | 10 | 9931 | XP_011538688.1 | |
| XM_011540387.2 | 6 | 11577 | XP_011538689.1 | |
| XM_017016969.1 | 15 | 8622 | XP_016872458.1 | |
| XM_017016970.1 | 14 | 8438 | XP_016872459.1 | |
| AK096575.1 | 1 | 220 | ||
| AK123570.1 | 1 | 1227 | ||
| AK295314.1 | ||||
| AL833070.2 | 1 | 2012 | CAD89969.1 | |
| BX647371.1 | 1 | 2625 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524966 | Chr.10:103441820 - 103717405 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map