Assay Details
Target Gene Details
Entrez Gene ID: | 220074 |
Gene Name: | leucine rich transmembrane and O-methyltransferase domain containing |
Gene Aliases: |
CFAP111, DFNB63, LRRC51 |
Location: |
Chr.11:72079982-72110782 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRTOMT | NM_001145307.4 | 1 | 147 | NP_001138779.1 |
| NM_001145308.4 | 1 | 147 | NP_001138780.1 | |
| NM_001145309.3 | 1 | 147 | NP_001138781.1 | |
| NM_001145310.3 | 1 | 147 | NP_001138782.1 | |
| NM_001205138.3 | 1 | 147 | NP_001192067.1 | |
| NM_001271471.2 | 1 | 147 | NP_001258400.1 | |
| NM_001318803.1 | 1 | 147 | NP_001305732.1 | |
| NM_145309.5 | 1 | 147 | NP_660352.1 | |
| NR_026886.3 | 1 | 147 | ||
| NR_134858.1 | 1 | 147 | ||
| XM_006718473.3 | 1 | 496 | XP_006718536.1 | |
| XM_006718474.3 | 1 | 496 | XP_006718537.1 | |
| XM_011544847.2 | 1 | 496 | XP_011543149.1 | |
| XM_011544848.2 | 1 | 496 | XP_011543150.1 | |
| XM_017017359.1 | 1 | 496 | XP_016872848.1 | |
| XM_017017360.1 | 1 | 496 | XP_016872849.1 | |
| DA151871.1 | 1 | 147 | ||
| DA637921.1 | 1 | 142 | ||
| DA664751.1 | 1 | 160 |
Target Gene Details
Entrez Gene ID: | 4926 |
Gene Name: | nuclear mitotic apparatus protein 1 |
Gene Aliases: |
NMP-22, NUMA |
Location: |
Chr.11:72002864-72080693 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUMA1 | NM_001286561.1 | NP_001273490.1 | ||
| NM_006185.3 | NP_006176.2 | |||
| NR_104476.1 | ||||
| XM_011545055.1 | XP_011543357.1 | |||
| XM_011545056.2 | XP_011543358.1 | |||
| XM_011545057.2 | XP_011543359.1 | |||
| XM_017017830.1 | XP_016873319.1 | |||
| AB210007.1 | ||||
| AB621824.1 | ||||
| AK307577.1 | ||||
| BC027493.1 | ||||
| BC043499.1 | AAH43499.1 | |||
| BC103765.1 | AAI03766.1 | |||
| BG771637.1 | ||||
| Z11583.1 | CAA77669.1 | |||
| Z11584.1 | CAA77670.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428262 | Chr.11:71652593 - 72093009 on Build GRCh38 | Gain+Loss |
|
| nsv384 | Chr.11:72074236 - 72106174 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map