Assay Details
Target Gene Details
Entrez Gene ID: | 9464 |
Gene Name: | heart and neural crest derivatives expressed 2 |
Gene Aliases: |
DHAND2, Hed, Thing2, bHLHa26, dHand |
Location: |
Chr.4:173526501-173530227 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HAND2 | NM_021973.2 | 2 | 1890 | NP_068808.1 |
| AK122739.1 | 3 | 1285 | ||
| BF313018.1 | ||||
| FJ226608.1 | 2 | 1892 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518998 | Chr.4:173501862 - 173532932 on Build GRCh38 | Loss |
|
| nsv522261 | Chr.4:173522345 - 173538672 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map