Assay Details
Target Gene Details
Entrez Gene ID: | 126147 |
Gene Name: | netrin 5 |
Gene Aliases: |
- |
Location: |
Chr.19:48661407-48673119 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NTN5 | NM_145807.2 | NP_665806.1 | ||
| XM_011526443.2 | XP_011524745.1 | |||
| XM_017026274.1 | XP_016881763.1 | |||
| BC018654.2 | AAH18654.2 | |||
| BC018697.2 | AAH18697.2 | |||
| BC021210.2 | 5 | 1410 | AAH21210.2 | |
| BC033207.1 | AAH33207.1 | |||
| BE270224.1 |
Target Gene Details
Entrez Gene ID: | 653677 |
Gene Name: | secretory blood group 1, pseudogene |
Gene Aliases: |
SEC1 |
Location: |
Chr.19:48638039-48682245 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEC1P | NR_004401.2 | |||
| AK301046.1 | ||||
| AK301238.1 | ||||
| D87946.1 | ||||
| D87947.1 | ||||
| DC353776.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map